Description
- Multiple holes to support 12 test tubes
- Dual handles easily secures holder in the ultrasonic cleaner
- Stainless steel
- Light and easy to maneuver
Specifications
Specifications
| Product Type | Test Tube Holder |
| For Use With (Equipment) | FB11211 Ultrasonic Cleaning Unit |
| Material | Stainless Steel |