Learn More
Fisherbrand™ Universal All Purpose Absorbent Loose Particulate
Catalog No. 19140917 Shop All Fisherbrand Products
Change view
Click to view available options
Material:
Polypropylene
Catalog No. Material
19-140-917 Polypropylene
1 options
Catalog No. 19-140-917
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Ideal for spill clean up, lab packing and stabilization of free liquids
  • Granular absorbents are eco-friendly and absorb oil, solvents, lubricants, coolants, water and most non-aggressive liquids on contact
  • Granular is a loose, multi-purpose absorbent ideal for liquid spill clean-up, lab packing and stabilization of free liquids
Compliance
OSHA 29 CFR 1910.22(a)(2); EPA 40 CFR 112.7(c)(1)(vii); OSHA 29 CFR 1910,120(j)(1)(vii).
Specifications
Color Gray
Material Polypropylene
Type Universal All-Purpose Absorbent Loose Particulate
Weight (English) 27 lb.
Weight (Metric) 12.2 kg