Learn More
Fisherbrand™ Urinalysis System
Catalog No. 14375246 Shop All Fisherbrand Products
Change view
Click to view available options
Disposable:
Single-use
Catalog No. Disposable
14-375-246 Single-use
1 options
This item is not returnable. View return policy
Catalog No. 14-375-246
Add to Cart
Add to Cart
This item is not returnable. View return policy
Includes
Includes 500 specimen collection cups in five bags of 100, plastic collection tubes, white polyethylene (plastic) caps, pressure-sensitive labels and five transport racks
For specimen collection and transport
All componenets are disposable for convenient collection, delivery and testing of urine specimens
  • Pressure-sensitive patient identification labels have ample writing space
  • Small, lightweight, easy-to-carry transport rack provides space for up to 20 tubes, labels and caps
  • Sturdy polystyrene tubes are centrifuge-safe
  • Tubes flare at the top to allow use of midget urinometer and ensure tight cap fit
Order Info
Tubes are available separately
Specifications
Product Type Urinalysis System
Disposable Single-use
For Use With (Application) For specimen collection and transport
Includes Flared tops, 500 Specimen collection cups, Plastic collection tubes, white Polyethylene caps, pressure-sensitive labels and five transport racks