Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Urine Cytology/FISH Collection and Transport Kit

Catalog No. 22130558 Shop All Fisherbrand Products
Change view
Click to view available options
Volume (Metric):
180 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
22-130-558 180 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-130-558

missing translation for 'validMainframeMsg'

Includes

  • Outer corrugated kit transport box (4.5 x 4.25 x 5 in.)
  • Secondary containment bag (8 x 8 in.)
  • Inner foam placeholder insert
  • 180mL Collection cup with 60mL minimum fill line label
  • 120mL Primary container transport jar† filled with 30mL of 70% reagent alcohol preservative
  • 3 oz. Refrigerant pack
  • Absorbent pad (5 x 7 in.)
  • UN3373 Biological Substance Category B label

Provides completely integrated collection and transport system for pathology-based laboratories

Kit includes all necessary components for for sample collection and transport — just add a common carrier air waybill and preprinted shipping envelope for return shipment of the kit to your laboratory

  • Ideal for laboratories expanding their test menu, geographic reach, or outreach testing programs
  • Outer corrugated kit box includes preprinted packing instructions
  • Kits are certified in compliance with DOT and IATA triple packaging requirements
  • Also compliant for ground and courier transport

Order Info

Shipped 16 per case

†Denotes 95kPa-rated primary container

Specifications

Product Type Urine Cytology Collection and Transport Kit
Certifications/Compliance DOT IATA
For Use With (Application) For pathology-based laboratories
Includes 4.5 x 4.25 x 5 in. Outer corrugated kit transport box, 8 x 8 in. secondary containment bag, inner foam placeholder insert, 180 mL collection cup with 60 mL minimum fill line label, 120 mL Primary container transport jar with 30 mL of 70% reagent alcohol preservative, 3 oz. refrigerant pack, 5 x 7 in. absorbent pad, UN3373 biological substance category B label
Volume (Metric) 180 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.