Learn More
Fisherbrand™ Viscometer Holders
SDP
Catalog No. 136179 Shop All Fisherbrand Products
Change view
Click to view available options
Material:
Neoprene, Rubber
Catalog No. Material
13-617-9 Neoprene, Rubber
1 options
Catalog No. 13-617-9
Add to Cart
Add to Cart
For use with Cannon-Manning Vacuum and Asphalt Institute Vacuum viscometers.
  • Available in either molded plastic or neoprene rubber to fit holes with a 2 in. diameter (5.1cm)
  • Plastic holders feature spring clip to hold viscometer securely
Specifications
Product Type Viscometer Holder
For Use With (Equipment) Cannon Manning Vacuum and Asphalt Institute Vacuum Viscometers
Material Neoprene, Rubber