Learn More
Fisherbrand™ Visitorspec Safety Glasses, Coated, Antifog
Catalog No. 191302091 Shop All Fisherbrand Products
Change view
Click to view available options
Lens Coating:
Anti-fog, Anti-static, Scratch-resistant
Catalog No. Lens Coating
19-130-2091 Anti-fog, Anti-static, Scratch-resistant
1 options
This item is not returnable. View return policy
Catalog No. 19-130-2091
Add to Cart
Add to Cart
This item is not returnable. View return policy
Ideal for protecting visitors in industrial or medical settings.
  • Economical safety spectacles
  • 100% impact-resistant polycarbonate wraparound unilens design with blue temples
  • Clear lens absorbs >99.9% of harmful UV light
  • Fit over prescription eyeglasses
  • Scratch-resistant, Anti-fog, Anti-static
Compliance
Meet ANSI™ Z87.1-2010 standards.
Specifications
Lens Coating Anti-fog, Anti-static, Scratch-resistant
Shape Rectangle
Certifications/Compliance Meets ANSI Z87.1-2003 standards.
Lens Tint Clear
Product Type Safety Glasses
Frame Color Clear
Temple Style Adjustable-length; spatula
Material Polycarbonate (Lens)
Lens Material Polycarbonate