Learn More
Fisherbrand™ Visitorspec Safety Glasses
SCI_ED
Catalog No. S01819 Shop All Fisherbrand Products
Change view
Click to view available options
Lens Coating:
Anti-fog, Anti-static, Scratch-resistant
Catalog No. Lens Coating
S01819 Anti-fog, Anti-static, Scratch-resistant
1 options
This item is not returnable. View return policy
Catalog No. S01819
This item is not returnable. View return policy
Ideal for protecting visitors in industrial or medical settings.
Economical safety spectacles available with coated or uncoated lens.
Uncoated Lens
  • 100% impact-resistant polycarbonate wraparound unilens design with blue temples
  • Clear lens absorbs >99.9% of harmful UV light
Coated Lens
  • 100% impact-resistant lens absorbs >99.9% of harmful UV light
  • Polycarbonate wraparound style with molded-in browguard and single lens
  • Coated to resist scratching, fogging and static
  • Adjustable-length spatula temples ensure a secure, comfortable fit
Compliance
Meet ANSI™ Z87.1-2003 standards.
Specifications
Lens Coating Anti-fog, Anti-static, Scratch-resistant
Certifications/Compliance Meet ANSI Z87.1-2003 standards.
Frame Color Blue
No. per Pack 12/Pk.
Temple Style Adjustable-length; spatula
Lens Material Polycarbonate