Learn More
Fisherbrand™ Visitorspec Safety Glasses
SCI_ED
Catalog No. p-3973426 Shop All Fisherbrand Products
Change view
Click to view available options
Lens Coating:
Anti-fog, Anti-static, Scratch-resistant
Uncoated
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Coating
S01819 Anti-fog, Anti-static, Scratch-resistant
S01820 Uncoated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. S01819
This item is not returnable. View return policy
Ideal for protecting visitors in industrial or medical settings.
Economical safety spectacles available with coated or uncoated lens.
Uncoated Lens
  • 100% impact-resistant polycarbonate wraparound unilens design with blue temples
  • Clear lens absorbs >99.9% of harmful UV light
Coated Lens
  • 100% impact-resistant lens absorbs >99.9% of harmful UV light
  • Polycarbonate wraparound style with molded-in browguard and single lens
  • Coated to resist scratching, fogging and static
  • Adjustable-length spatula temples ensure a secure, comfortable fit
Compliance
Meet ANSI™ Z87.1-2003 standards.
Specifications
Lens Coating Anti-fog, Anti-static, Scratch-resistant
Certifications/Compliance Meet ANSI Z87.1-2003 standards.
Frame Color Blue
No. per Pack 12/Pk.
Temple Style Adjustable-length; spatula
Lens Material Polycarbonate