Learn More
Fisherbrand™ White Line Flasks
SDP
Catalog No. p-126315 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (Metric):
150 mL
Catalog No. Capacity (Metric)
10-200J 150 mL
1 options
Catalog No. 10-200J
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
Single graduation ring blasted on the neck.
Same as 10-205 Series, except without stoppers and ground glass tops. Use rubber stoppers.
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 150 mL