Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Dry Transport Swab

Catalog No. 1490711 Shop All Fisherbrand Products
Change view
Click to view available options
Swab Configuration:
Dual Applicator
Single Applicator
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Swab Configuration
14-907-11 Dual Applicator
14-907-10 Single Applicator
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 14-907-11
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Fisherbrand™ Dry Transport Swab features a single or double rayon swab with plastic shaft in a spiral, round bottom tube with white plug cap. Swabs are provided sterile.

 

  • Dry swab
  • Single or double swab
  • Patient identification label on swab tube
  • Lot number and expiration date printed on tube labels
  • Swabs are tipped with inert, nontoxic rayon
  • White caps
  • Gamma radiation sterilized
  • Shelf life of 3 years
Note: There is no media in casing for dry swab to transport a sample, esp. those of aerobic or anaerobic state.

 

Specifications

Product Type Transport Swab
Format Swab
Sterility Sterile
Swab Configuration Dual Applicator
Closure Color White
For Use With (Application) For the collection and/or transportation of a wide range of bacteria, viruses and facultative microorganisms
Packaging Peel pouch
Quantity 100/Pk.
Shaft Material Plastic
Tip Material Rayon
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.