Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FisherTab™ TT-57 Kjeldahl Tablets, Fisher Chemical

Catalog No. K3181000 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1000 Tablets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K3181000 1000 Tablets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K3181000

missing translation for 'validMainframeMsg'

Composition per tablet: 5.57g K2SO4 + 0.0033g CuSO4 + 0.2g TiO2

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material FisherTab™ TT-57 Kjeldahl Tablets
Color Blue
CAS 13463-67-7, 7778-80-5, 7758-98-7
Physical Form Powder/Solid
Quantity 1000 Tablets
Assay Percent Range 96%
Packaging Poly Bottle
MDL Number MFCD00010981
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.