Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Fishing Line for Science Kits
SCI_ED

Catalog No. S05191 Shop All Fisherbrand Products
Change view
Click to view available options
:
10 lb. test; 650 yd.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S05191 10 lb. test; 650 yd.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S05191

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Consumable for use with multiple projects and with science kits.

Specifications

Product Type Fishing Line String
For Use With (Application) For multiple projects or in a science kit
Length (English) 650 yd.
Length (Metric) 594.3 m
Weight (English) 10 lb.
Weight (Metric) 4.5 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.