Learn More
Selleck Chemical LLC Fisogatinib
SDP
Supplier:  Selleck Chemical LLC S850325MG
Encompass_Preferred
Fisogatinib (BLU-554) is a potent highly-selective oral FGFR4 inhibitor with an IC50 value of 5 nM The IC50s for FGFR1-3 is 624-2203 nM
Catalog No. 50-000-00658