Learn More
LabStrong Corp FISTREEM HEAT ELEMENT 120V 20A
Catalog No. NC0020978
Encompass_Preferred
LabStrong part number EL744X3LS compares to Fi-Streem part number EL744X3. The heating element (120V, 900W, 20A Stills) is a replacement part for Fi-Streem Water Purification Stills. The heating element includes the plastic element union and heater sheath.
Catalog No. NC0020978 Supplier LabStrong Corp Supplier No. EL744X3LS
LabStrong part number EL744X3LS compares to Fi-Streem part number EL744X3. The heating element (120V, 900W, 20A Stills) is a replacement part for Fi-Streem Water Purification Stills. The heating element includes the plastic element union and heater sheath.