Learn More
Dot Scientific FIT PIPET TIP .1-10UL 960/PK
SDP
Catalog No. NC1662062
Encompass
BioDot Universal Fit Pipet Tip .1-10ul Extended Length Pipetman Style Low Retention Graduated Beveled Natural Sterile 96 Tips Per Cell Rack 960 Tips Per Pack
Catalog No. NC1662062 Supplier Dot Scientific Supplier No. RN0P10XLNBCRS
BioDot Universal Fit Pipet Tip .1-10ul Extended Length Pipetman Style Low Retention Graduated Beveled Natural Sterile 96 Tips Per Cell Rack 960 Tips Per Pack