Learn More
Ibidi USA Fitting Reducer 0.5/0.8 to 1.6
SDP
Catalog No. NC1515755
Encompass
Fitting reducer for adapting 0.5 or 0.8 mm ID tubing to 1.6 mm ID silicone tubing. polypropylene, sterile, 25 pcs./box
This item is not returnable. View return policy
Catalog No. NC1515755 Supplier Ibidi USA Supplier No. 10829
Fitting reducer for adapting 0.5 or 0.8 mm ID tubing to 1.6 mm ID silicone tubing. polypropylene, sterile, 25 pcs./box