Learn More
Med Vet International 3M QualitatIVe Fit Test Kit, Sweet, Ft-10, 1 Kit
Catalog No. 502830774
Encompass_Preferred
The 3M™ fit test kit includes two nebulizers, a hood and collar, a sweet sensitivity solution and a sweet-tasting test solution to supply you with a full kit It ensures the seal on any particulate or gas respirator is functioning properly The included test solution has sodium saccharin to produce a sweet-tasting aerosol The 3M™ qualitative fit test makes it easy to determine performance efficiency It meets performance criteria for fit testing respirators per current OSHA rules
Catalog No. 50-283-0774 Supplier Med Vet International Supplier No. 230067KT
The 3M™ fit test kit includes two nebulizers, a hood and collar, a sweet sensitivity solution and a sweet-tasting test solution to supply you with a full kit It ensures the seal on any particulate or gas respirator is functioning properly The included test solution has sodium saccharin to produce a sweet-tasting aerosol The 3M™ qualitative fit test makes it easy to determine performance efficiency It meets performance criteria for fit testing respirators per current OSHA rules