Learn More
Dentec Safety™ Bitrex Fit Test Kit
SDP
Catalog No. 19222545 Shop All Dentec Safety Specialists Inc. Products
Change view
Click to view available options
For Use With (Equipment):
Masks
Catalog No. For Use With (Equipment)
19-222-545 Masks
1 options
Catalog No. 19-222-545 Supplier Dentec Safety™ Supplier No. 79260
Add to Cart
Add to Cart
Fit test kit includes Bitrex, test hood, nebulizer 1, nebulizer 2, sensitivity solution, fit test solution, and 2 sets of replacement nebulizer inserts.
  • Includes fit test hood, nebulizer #1, nebulizer #2, sensitivity solution, fit test solution, and two sets of replacement nebulizer inserts
Specifications
For Use With (Equipment) Masks
Model 79260