Learn More
Dentec Safety™ Banana Oil Fit Test Kit
SDP
Catalog No. 19222543 Shop All Dentec Safety Specialists Inc. Products
Change view
Click to view available options
For Use With (Equipment):
Masks
Catalog No. For Use With (Equipment)
19-222-543 Masks
1 options
Catalog No. 19-222-543 Supplier Dentec Safety™ Supplier No. 79240
Add to Cart
Add to Cart
Fit Test Kit includes Banana Oil, (2) OV Cartridges, and instructions.
  • Fit testing kit
  • Includes banana oil two organic vapor cartridges
  • Meets the NIOSH standard
Specifications
For Use With (Equipment) Masks
Model 79240