Learn More
Epigentek Inc FitAmp Blood and Cultured Cell DNA Extraction Kit (50 samples)
SDP
Catalog No. NC0706313
Encompass_Preferred
The FitAmp Blood and Cultured Cell DNA Extraction Kit is a complete set of essential components which enables the experimenter to efficiently isolate DNA from blood leukocytes and cultured mammalian cells. The entire procedure can be completed within 20 minutes.
Catalog No. NC0706313 Supplier Epigentek Inc Supplier No. P1018050
The FitAmp Blood and Cultured Cell DNA Extraction Kit is a complete set of essential components which enables the experimenter to efficiently isolate DNA from blood leukocytes and cultured mammalian cells. The entire procedure can be completed within 20 minutes.