Learn More
Medchemexpress LLC HY-66019 100mg Medchemexpress, FITC CAS:3326-32-7 Purity:>98%
SDP
Catalog No. 502030958
Encompass_Preferred
Medchemexpress, HY-66019 100mg FITC CAS:3326-32-7 FITC is a derivative of fluorescein for the labeling of amines. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0958 Supplier Medchemexpress LLC Supplier No. HY66019100MG
Medchemexpress, HY-66019 100mg FITC CAS:3326-32-7 FITC is a derivative of fluorescein for the labeling of amines. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.