Learn More
Medchemexpress LLC HY-66019 500mg , FITC CAS:3326-32-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY66019500MG
Encompass_Preferred
HY-66019 500mg FITC CAS: 3326-32-7 FITC (Fluorescein 5-isothiocyanate) is a fluorescence probe for the labeling of amines. FITC is a pH- and Cu2+-sensitive fluorescence dye. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0265