Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC FITC-β-Amyloid (25-35) | 99.4% | 1562.81 | 5 MG
SDP

Catalog No. 5000426177
Encompass_Preferred

FITC-β-Amyloid (25-35) is a FITC-labeled β-Amyloid (25-35) peptide, developed for research. It is relevant to the study of neurological diseases, particularly neurodegenerative conditions like Alzheimer's Disease, and is associated with neuronal signaling and Amyloid-β research.

  • Purity: 99.4%
  • Molecular weight: 1562.81
  • Formula: C72H103N15O20S2
  • Appearance: solid
  • Color: yellow to orange
  • Sequence: FITC-Ahx-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met (shortened as FITC-Ahx-GSNKGAIIGLM)
  • Storage: powder -80°C for 2 years or -20°C for 1 year; in solvent -80°C for 6 months or -20°C for 1 month (sealed, away from moisture).

Catalog No. 50-004-26177 Supplier Medchemexpress LLC Supplier No. HYP0128F5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

FITC-β-Amyloid (25-35) is a FITC-labeled β-Amyloid (25-35) peptide, developed for research. It is relevant to the study of neurological diseases, particularly neurodegenerative conditions like Alzheimer's Disease, and is associated with neuronal signaling and Amyloid-β research.

  • Purity: 99.4%
  • Molecular weight: 1562.81
  • Formula: C72H103N15O20S2
  • Appearance: solid
  • Color: yellow to orange
  • Sequence: FITC-Ahx-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met (shortened as FITC-Ahx-GSNKGAIIGLM)
  • Storage: powder -80°C for 2 years or -20°C for 1 year; in solvent -80°C for 6 months or -20°C for 1 month (sealed, away from moisture).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.