Learn More
Cosmo Bio Usa Inc FITC Anti Collagen 4TX Red, 1 ml, Monoclonal Antibody Cocktail, Rat, Clones H53, B51, H25, Immunofluorescence
SDP
Supplier:  Cosmo Bio Usa Inc SGECFT45325
Encompass
The COSMO BIO USA INC SGE-CFT45325 is a specialized monoclonal antibody cocktail designed for immunofluorescent analysis, particularly in diagnosing Alport syndrome through the identification of specific collagen chains. This solution contains fluorochrome-conjugated antibodies providing rapid and effective localization of collagen within human renal and skin biopsies.
  • Monoclonal antibody cocktail
  • Designed for immunofluorescence applications
  • Targets human collagen α5(IV) and α2(IV)
  • Includes two FITC-conjugated and one Texas Red-conjugated antibody
  • Developed for quick staining of cryostat sections
  • Stored in the dark at 2-4°C
  • Stability of 2 years under proper conditions
  • Research use only
Catalog No. NC1112065