Learn More
Biolegend FITC anti-human CD326 (EpCAM)
Supplier:  Biolegend 369814
Encompass_Preferred
FITC anti-human CD326 (EpCAM) [CO17-1A]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. NC1295864