Learn More
Biolegend FITC anti-human CD104
Supplier:  Biolegend 327805
Encompass_Preferred
FITC anti-human CD104 [58XB4]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-167-386