Learn More
Biolegend FITC anti-human CD107a (LAMP-1)
Supplier:  Biolegend 328606
Encompass_Preferred
FITC anti-human CD107a (LAMP-1) [H4A3]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Chimpanzee, Cynomolgus, Pigtailed Macaque, Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-167-455