Learn More
Biolegend FITC anti-human CD11c
Supplier:  Biolegend 337214
Encompass_Preferred
FITC anti-human CD11c [Bu15]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-167-902