Learn More
Biolegend FITC anti-human CD158b (KIR2DL2/L3, NKAT2)
Supplier:  Biolegend 312603
Encompass_Preferred
FITC anti-human CD158b (KIR2DL2/L3, NKAT2) [DX27]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-166-687