Learn More
Biolegend FITC anti-human CD158e1 (KIR3DL1, NKB1)
Supplier:  Biolegend 312705
Encompass_Preferred
FITC anti-human CD158e1 (KIR3DL1, NKB1) [DX9]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-166-693