Learn More
Biolegend FITC anti-human CD177
Supplier:  Biolegend 315804
Encompass_Preferred
FITC anti-human CD177 [MEM-166]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-166-843