Learn More
Biolegend FITC anti-human CD203c (E-NPP3)
Supplier:  Biolegend 324613
Encompass_Preferred
FITC anti-human CD203c (E-NPP3) [NP4D6]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-207-5670