Learn More
Biolegend FITC anti-human CD235a (Glycophorin A)
Supplier:  Biolegend 349103
Encompass_Preferred
FITC anti-human CD235a (Glycophorin A) [HI264]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 25 ug
Catalog No. 50-605-298