Learn More
Biolegend FITC anti-human CD268 (BAFF-R)
Supplier:  Biolegend 316904
Encompass_Preferred
FITC anti-human CD268 (BAFF-R) [11C1]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-166-890