Learn More
Biolegend FITC anti-human CD271 (NGFR)
Supplier:  Biolegend 345104
Encompass_Preferred
FITC anti-human CD271 (NGFR) [ME20.4]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-168-330