Learn More
Biolegend FITC anti-human CD282 (TLR2)
Supplier:  Biolegend 309706
Encompass_Preferred
FITC anti-human CD282 (TLR2) [TL2.1]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-166-514