Learn More
Biolegend FITC anti-human CD3
Supplier:  Biolegend 317306
Encompass_Preferred
FITC anti-human CD3 [OKT3]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-166-907