Learn More
Biolegend FITC anti-human CD3
Supplier:  Biolegend 981002
Encompass_Preferred
FITC anti-human CD3 [SK7]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 500 uL
Catalog No. 50-207-5505