Learn More
Biolegend FITC anti-human CD3
Supplier:  Biolegend 300440
Encompass_Preferred
FITC anti-human CD3 [UCHT1]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee; Apps: FC; Size: 500 tests
Catalog No. 50-603-969