Learn More
Biolegend FITC anti-human CD303 (BDCA-2)
Supplier:  Biolegend 354208
Encompass_Preferred
FITC anti-human CD303 (BDCA-2) [201A]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-403-484