Learn More
Biolegend FITC anti-human CD318 (CDCP1)
Supplier:  Biolegend 324004
Encompass_Preferred
FITC anti-human CD318 (CDCP1) [CUB1]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-167-213