Learn More
Biolegend FITC anti-human CD4
Supplier:  Biolegend 317408
Encompass_Preferred
FITC anti-human CD4 [OKT4]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. NC9997971