Learn More
Biolegend FITC anti-human CD40
Supplier:  Biolegend 334305
Encompass_Preferred
FITC anti-human CD40 [5C3]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Baboon, Chimpanzee, Cynomolgus Monkey, Rhesus, Squirrel Monkey ; Apps: FC; Size: 25 tests
Catalog No. 50-167-771