Learn More
Biolegend FITC anti-human CD41
Supplier:  Biolegend 303703
Encompass_Preferred
FITC anti-human CD41 [HIP8]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Capuchin Monkey, Cynomolgus, Rhesus; Apps: FC; Size: 25 tests
Catalog No. 50-166-025