Learn More
Biolegend FITC anti-human CD42b
Supplier:  Biolegend 303903
Encompass_Preferred
FITC anti-human CD42b [HIP1]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee; Apps: FC; Size: 25 tests
Catalog No. 50-166-041