Learn More
Biolegend FITC anti-human CD45
Supplier:  Biolegend 304054
Encompass_Preferred
FITC anti-human CD45 [HI30]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee; Apps: FC; Size: 100 ug
Catalog No. 50-103-0637