Learn More
Biolegend FITC anti-human CD49e
Supplier:  Biolegend 328007
Encompass_Preferred
FITC anti-human CD49e [NKI-SAM-1]; Isotype: Mouse IgG2b, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 25 tests
Catalog No. 50-167-403