Learn More
Biolegend FITC anti-human CD52
Supplier:  Biolegend 316003
Encompass_Preferred
FITC anti-human CD52 [HI186]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Cynomolgus, Rhesus; Apps: FC; Size: 25 tests
Catalog No. 50-166-845