Learn More
Biolegend FITC anti-human CD59
Supplier:  Biolegend 304706
Encompass_Preferred
FITC anti-human CD59 [p282 (H19)]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Baboon, Chimpanzee, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-166-153