Learn More
Biolegend FITC anti-human CD79a (Igα)
Supplier:  Biolegend 333511
Encompass_Preferred
FITC anti-human CD79a (Igα) [HM47]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ICFC; Size: 25 tests
Catalog No. 50-207-6103